Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 23435 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-55376. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43 TAR DNA-binding protein 43 |
| Gene Symbol | TARDBP |
| Product Type | TDP-43/TARDBP |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
