Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TRIM5 alpha Antibody Blocking Peptide
SDP

Catalog No. NBP176600E Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176600E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176600E Supplier R&D Systems™ Supplier No. NBP176600PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

15 amino acids from the carboxy terminus of rhesus monkey TRIM5alpha. Source: Synthetic (Accession #: ABG67967) The TRIM5 alpha Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 85363
Species Rhesus Macaque
Product Line Novus
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76600. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Gene Symbol TRIM5
Product Type TRIM5 alpha
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Termination 15 amino acids from the carboxy terminus of rhesus monkey TRIM5alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.