Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ UDP glucose dehydrogenase Protein
SDP

Catalog No. NBP14527605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14527605 0.05 mg
NBP14527602 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14527605 Supplier Novus Biologicals™ Supplier No. NBP1452760.05MG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-494 of Human UDP glucose dehydrogenase The Recombinant Human UDP glucose dehydrogenase Protein is derived from E. coli. The Recombinant Human UDP glucose dehydrogenase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1mM DTT, 0.1M NaCl, and 1mM EDTA.
Gene ID (Entrez) 7358
Molecular Weight (g/mol) 59.5kDa
Purification Method Protein
Quantity 0.05 mg
Source Human
Immunogen MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSELEM FEIKKICCIG AGYVGGPTCS VIAHMCPEIR VTVVDVNESR INAWNSPTLP IYEPGLKEVV ESCRGKNLFF STNIDDAIKE ADLVFISVNT PTKTYGMGKG RAADLKYIEA CARRIVQNSN GYKIVTEKST VPVRAAESIR RIFDANTKPN LNLQVLSNPE FLAEGTAIKD LKNPDRVLIG GDETPEGQRA VQALCAVYEH WVPREKILTT NTWSSELSKL AANAFLAQRI SSINSISALC EATGADVEEV ATAIGMDQRI GNKFLKASVG FGGSCFQKDV LNLVYLCEAL NLPEVARYWQ QVIDMNDYQR RRFASRIIDS LFNTVTDKKI AILGFAFKKD TGDTRESSSI YISKYLMDEG AHLHIYDPKV PREQIVVDLS HPGVSEDDQV SRLVTISKDP YEACDGAHAV VICTEWDMFK ELDYERIHKK MLKPAFIFDG RRVLDGLHNE LQTIGFQIET IGKKVSSKRI PYAPSGEIPK FSLQDPPNKK PKV
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name UDP glucose dehydrogenase
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.