Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 6572 |
| Species | Human |
| Content And Storage | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| Formulation | Soluble in PBS and Water |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-91348. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter |
| Gene Symbol | SLC18A3 |
| Product Type | VAChT/SLC18A3 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
