Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant E. coli MYH His Protein
SDP

Catalog No. NBP222653 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222653 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222653 Supplier Novus Biologicals™ Supplier No. NBP222653
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-350 of E.coli MYH The Recombinant E. coli MYH Protein is derived from E. coli. The Recombinant E. coli MYH Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol
Gene ID (Entrez) 4595
Molecular Weight (g/mol) M.W. (theoretical): 41.5 kDa
Name MYH Protein
Purification Method SDS-PAGE
Quantity 0.01 mg
Source E. Coli
Immunogen mutY, 1-350aa, E.coli, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMQASQFS AQVLDWYDKY GRKTLPWQID KTPYKVWLSE VMLQQTQVAT VIPYFERFMA RFPTVTDLAN APLDEVLHLW TGLGYYARAR NLHKAAQQVA TLHGGKFPET FEEVAALPGV GRSTAGAILS LSLGKHFPIL DGNVKRVLAR CYAVSGWPGK KEVENKLWSL SEQVTPAVGV ERFNQAMMDL GAMICTRSKP KCSLCPLQNG CIAAANNSWA LYPGKKPKQT LPERTGYFLL LQHEDEVLLA QRPPSGLWGG LYCFPQFADE ESLRQWLAQR QIAADNLTQL TAFRHTFSHF HLDIVPMWLP VSSFTGCMDE GNALWYNLAQ PPSVGLAAPV ERLLQQLRTG APV
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol MUTYH
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Research Category Base Excision Repair, Cancer, DNA Repair
Purity or Quality Grade >90%, by SDS-PAGE
Protein MYH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.