Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 65125 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 600-225. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, Erythrocyte 65 kDa protein, HSAN2hereditary sensory neuropathy, type II, HSN2MGC163339, hWNK1, KDPMGC163341, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1PSK, prostate-derived sterile 20-like kinase, Protein kinase lysine-deficient 1, Protein kinase with no lysine 1, protein kinase, lysine deficient 1, serine/threonine-protein kinase WNK1, WNK lysine deficient protein kinase 1 |
| Gene Symbol | WNK1 |
| Product Type | WNK1 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
