Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Chromata ChIP Histone H3 [ac Lys4] Kit (ChIP)
SDP

Catalog No. nbp171710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 Immunoprecipitations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171710 25 Immunoprecipitations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171710 Supplier Novus Biologicals™ Supplier No. NBP171710
Only null left
Add to Cart
Add to Cart

Includes

10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair

Applications: Chromatin Immunoprecipitation

Kit contains all the necessary components to perform 25 Immunoprecipitation experiments including a positive control antibody. This ChIP kit is specific for Histone H3 acetylated Lysine 4.

Specifications

Kit Contents 10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair
Content And Storage Storage of components varies. See protocol for specific instructions.
Quantity 25 Immunoprecipitations
For Use With (Application) Chromatin Immunoprecipitation of DNA from fixed Hela cells alongside a no antibody (No Ab) control
Target Chromata ChIP Histone H3
Type ChIP Histone H3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.