Learn More
Description
- A blocking peptide containing 15 amino acids near the amino terminus of human NOX3
- NOX3 peptide is used for blocking the NOX3 antibody, NBP1-41292.
- Source: Synthetic
- Type: Blocking Peptide
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| Gene ID (Entrez) | 50508 |
| Protein Family | NOX3 |
| Concentration | 0.2 mg/mL |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | NOX3 |
| Quantity | 0.05 mg |
| Source | Synthetic |
| Storage Buffer | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
