Learn More
Boston Bioproducts Inc NP-40 Lysis Buffer - 500ml
Supplier:  Boston Bioproducts Inc BP119500ML
Encompass_Preferred
The NP-40 Lysis Buffer is a solubilizing agent used to prepare cell extracts for immunoassays and blotting techniques. As one of the most popular lysis buffers on the market, the NP-40 lysis buffers can support a variety of laboratory workflows. Relying on the non-ionic detergent NP-40 as its primary solubilizing agent, this Lysis buffer is weaker than a RIPA buffer, and can permeabilize cell membranes, partially open the nucleus without fully disrupting it, and lyse mitochondiral membranes.
Catalog No. NC9540077
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.