Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NP-40 lysis buffer, low salt, Thermo Scientific™

Catalog No. AAJ60902AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60902AK 250 mL
AAJ60902AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60902AK Supplier Thermo Scientific Chemicals Supplier No. J60902AK
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Lysis buffer for extracting proteins from cells. NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay, ELISA. This lysis buffer can be used to prepare cell lysates for western blots or immunoprecipitation. It can also be used as a wash buffer for immunoprecipitation reactions.

Solubility
Fully misicible in water.

Notes
This buffer contains: 50mM Tris-HCl (pH 7.4), 1% NP-40, and 5mM EDTA. To use, incubate cells in 5 volumes of buffer on ice for 10-30 minutes.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NP-40 lysis buffer
Name Note low salt
Color Colorless
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully misicible in water.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.