Learn More
Apexbio Technology LLC Avanafil 330784-47-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A43345.1
Encompass_Preferred
Avanafil (CAS 330784-47-9) is a potent and highly selective inhibitor of phosphodiesterase type 5 (PDE5) with an IC50 value of 5 2 nM and a Ki of 4 3 nM PDE5 is abundantly expressed in the corpus cavernosum where it modulates blood flow by regulating the hydrolysis of cGMP Avanafil acts by competitively binding to PDE5 thereby preventing cGMP breakdown and promoting vasodilation In preclinical studies avanafil enhanced penile erection in dose-dependent manners with ED200% values of 37 5 g/kg (intravenous plasma concentration 59 6 ng/ml) and 151 7 g/kg (duodenal) Avanafil is primarily utilized in research related to erectile dysfunction and vascular signal transduction
Catalog No. 50-000-93751
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.