Learn More
MBL International Corporation Anti-Np95 (Mouse) mAb (Monoclonal Antibody)
SDP
Supplier:  MBL International Corporation D2893
Encompass_Preferred
Clonality:Monoclonal; Application:FCM|ICC|IHC|IP|WB; Conjugate:Unlabeled; Species Reactivity:Mouse; Size:100 µl; Target:NP95; Np95 recognizes hemimethylated CpGs via the SRA domain, and recruits Dnmt1 to the sites to transfer the methylation patterns onto the newly synthesized DNA strand. Therefore, the interaction between Np95 and Dnmt1 is necessary for the inheritance of DNA methylation.Monoclonal antibody targets NP95 for FCM, ICC, IHC, IPP, WB.NP95 Monoclonal Antibody.
Catalog No. 50-461-69
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.