Learn More
Invitrogen™ NPAS4 Monoclonal Antibody (N408/79), APC

Description
A 1:100 dilution of MA5-45749 was sufficient for detection of NPAS4 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 90kDa.approximately This antibody was formerly sold as clone S408-79.
Specifications
Specifications
| Antigen | NPAS4 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N408/79 |
| Concentration | 1 mg/mL |
| Conjugate | APC |
| Formulation | 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 |
| Gene | NPAS4 |
| Gene Accession No. | Q8BGD7, Q8CJH6, Q8IUM7 |
| Gene Alias | bHLH PAS type transcription factor NXF; bHLHe79; bHLH-PAS type transcription factor NXF; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; neuronal PAS domain protein 4; neuronal PAS domain-containing protein 4; Neuronal PAS4; Npas4; NXF; PAS domain-containing protein 10; PASD10; PASD10 and LE-PAS |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.