Learn More
Enzo Life Sciences Nuclear pore complex protein Nup153 monoclonal antibody (QE5) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1110100
Encompass_Preferred
Clone: QE5. Isotype: IgG1. Immunogen: Rat liver nuclear envelope proteins. Species reactivity: Eukaryote. Specificity: Recognizes eukaryote NUP153 as well as two related nuclear pore complex proteins: NUP214 and p62. By immunofluorescence, QE5 labels the nuclear envelope of eukaryotic cells giving a punctate staining pattern. Applications: IF, IP, WB. Purity: Purified. Formulation: Liquid. In PBS containing 0.03% thimerosal.
Catalog No. 50-201-2668
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.