Learn More
Invitrogen™ NPM1 Monoclonal Antibody (FC-61991), Alexa Fluor™ 555
Shop All Thermo Scientific Products

Description
NPM1 Monoclonal Antibody for ICC/IF
Specifications
Specifications
| Antigen | NPM1 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | FC-61991 |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 555 |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene | NPM1 |
| Gene Accession No. | P06748, P13084, Q61937 |
| Gene Alias | B23; B23 antibody; B23NP; MGC104254; mNPM antibody; mNPM1 antibody; NO38; NO38 antibody; Npm; npm1; npm1.L; Nucleolar phosphoprotein B23; nucleolar protein B23.1; nucleolar protein B23/No38; nucleolar protein NO38; Nucleophosmin; nucleophosmin (nucleolar phosphoprotein B23, numatrin); nucleophosmin (nucleolar phosphoprotein B23, numatrin) L homeolog; nucleophosmin 1; nucleophosmin/nucleoplasmin family, member 1; nucleoplasmin; numatrin; numatrin antibody; testicular tissue protein Li 128; unnamed protein product; XELAEV_18020022mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.