Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPRL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578244 Supplier Invitrogen™ Supplier No. PA578244
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: K562, U87-MG, SK-N-SH Predicted Reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The function of the encoded protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRL3
Applications Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NPRL3
Gene Accession No. Q12980
Gene Alias -14 gene protein; Aag; alpha-globin regulatory element-containing gene protein; alphaRE-14; C16orf35; CGTHBA; conserved gene telomeric to alpha globin cluster; GATOR complex protein NPRL3; HS-26; HS40; HS-40; MARE; nitrogen permease regulator 3-like protein; NPR3; NPR3 like, GATOR1 complex subunit; NPR3-like; NPR3-like, GATOR1 complex subunit; NPRL3; Phg; Protein CGTHBA; Prox1; RMD11; RP23-177M10.6
Gene Symbols NPRL3
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Human NPRL3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.