Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3212 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3212 Supplier Invitrogen™ Supplier No. PA3212
  • $576.80
    $449.65 / Each of 1

    Save $127.15

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-212 detects NPS in human, mouse, and rat samples. PA3-212 has successfully been used in immunocytochemical, immunofluorescence and immunohistochemical (paraffin) procedures. The PA3-212 immunogen is a synthetic peptide corresponding to SFRNGVGTGMKKTSFQRAKS of NPS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NPS
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.02% sodium azide
Gene NPS
Gene Accession No. P0C0P6, P0C0P7, P0C0P8
Gene Alias ENSMUSG00000073804; Neuropeptide S; NPS; prepro-neuropeptide S
Gene Symbols NPS
Host Species Rabbit
Immunogen Synthetic peptide corresponding to SFRNGVGTGMKKTSFQRAKS of NPS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100043254, 100360071, 594857
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.