Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTI NPX Series White Nitrile Gloves

Catalog No. 19050346
Change view
Click to view available options
Size:
Small
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-346 2X-Large
19-050-342 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-050-346 Supplier CTI Supplier No. NPX105
This item is not returnable. View return policy

White, powder free nitrile gloves feature textured finger tips with a soft feel.

  • White nitrile
  • Powder free
  • Textured finger tips and palm
  • 10 in. Length
  • Ambidextrous
  • Tapered, Beaded Cuff
  • Latex-free

Specifications

Certifications/Compliance ASTM D 3578-95 Standards, MIL STD-105E, 21 CFR 177-2600
Chemotherapeutic Rated No
Applications Laboratory, Healthcare, Industrial
Color White
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 2X-Large
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
FDA 510 (k) Yes
Model NPX105
Series NPX
Tensile Strength 14 MPa
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.