Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA124891
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA124891 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA124891 Supplier Invitrogen™ Supplier No. PA124891
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: rat brain. PA1-24891 is expected to cross react with chicken (99% conserved) due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a neuropeptide that is widely expressed in the central nervous system and influences many physiological processes, including cortical excitability, stress response, food intake, circadian rhythms, and cardiovascular function. The neuropeptide functions through G protein-coupled receptors to inhibit adenylyl cyclase, activate mitogen-activated protein kinase, regulate intracellular calcium levels, and activate potassium channels. A polymorphism in this gene resulting in a change of leucine 7 to proline in the signal peptide is associated with elevated cholesterol levels, higher alcohol consumption, and may be a risk factor for various metabolic and cardiovascular diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY
Applications ELISA, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 15mM sodium azide
Gene NPY
Gene Accession No. P01303, P01304, P07808, P14765, P57774
Gene Alias 0710005A05Rik; C-flanking peptide of NPY; CPON; involved in appetite regulation; LOC100912228; Neuropeptide tyrosine; Neuropeptide Y; neuropeptide Y precursor; Neuropeptide-Y; Npy; NPY02; OTTHUMP00000201947; prepro-neuropeptide Y; proneuropeptide Y; pro-neuropeptide Y; pro-neuropeptide Y-like; PYY4; RATNPY; RATNPY02
Gene Symbols NPY
Host Species Rabbit
Immunogen Synthetic neuropeptide Y (NPY, porcine), conjugated to KLH.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100912228, 109648, 24604, 397304, 443508, 4852
Target Species Human, Mouse, Rat, Sheep, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.