Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPY1R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106849 Supplier Thermo Scientific Supplier No. PA5106849
Only null left
Add to Cart
Add to Cart

NPY1R Polyclonal Antibody

NPY1R Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY1R
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NPY1R
Gene Accession No. P21555, P25929, Q04573
Gene Alias FC5; Neuropepetide Y1 Receptor; neuropeptide Y receptor subtype; neuropeptide Y receptor type 1; neuropeptide Y receptor Y1; neuropeptide Y1 receptor; NPY-1; NPY-1 receptor; Npy1r; NPY1-R; NPYR; NPYY1; NPY-Y1 receptor; PY receptor Y1; Y1-R
Gene Symbols NPY1R
Host Species Rabbit
Immunogen A synthesized peptide derived from human NPY1R(Accession P25929), corresponding to amino acid residues F221-A271.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18166, 29358, 4886
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.