Learn More
NQO2 Mouse anti-Canine, Human, Clone: OTI1G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504784

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NQO2 (EC 1.10.99.2) is a flavoprotein that catalyzes the 2-electron reduction of various quinones, redox dyes, and the vitamin K menadione. NQO2 predomitly uses dihydronicotinamide riboside (NRH) as the electron donor.Specifications
NQO2 | |
Monoclonal | |
Unconjugated | |
NQO2 | |
DHQV, DIA6, NMOR2, QR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4835, 606932 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P16083 | |
NQO2 | |
Full length human recombit protein of human NQO2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.