Learn More
OTT Hydromet Kipp & Zonen - NR LITE 2 RADIOMETER
Supplier:  OTT Hydromet 0344920003
Encompass_Preferred
NR Lite2 is designed for routine measurement of net radiation which is the balance between incoming and outgoing radiation under outdoor conditions. The design of the NR Lite2 is unique. The double-sided detector has black conical absorbers with an anti-stick weather resistant protective coating. In contrast to other sensor designs, NR Lite2 requires no fragile plastic domes. This results in a virtually maintenance free design. The vertical stick prevents birds from affecting the output signal. The NR Lite2 is easy to use. It is based on a thermopile sensor. The voltage is proportional to the net radiation. It can be directly connected to voltmeter or data logger with an mV input. It has the same high quality yellow cable fitted as our other instruments. The NR Lite2 is suitable for outdoor use conditions and fully complies with CE regulations.
Catalog No. NC1919648
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.