Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2F1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7210466 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4430 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4430 Supplier Proteintech Group Inc Supplier No. 245731AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a nuclear hormone receptor and transcriptional regulator. The encoded protein acts as a homodimer and binds to 5′-AGGTCA-3′ repeats. Defects in this gene are a cause of Bosch-Boonstra optic atrophy syndrome (BBOAS).

Specifications

Antigen NR2F1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NR2F1
Gene Accession No. P10589, Q60632
Gene Alias COUP TF I, COUP TF1, COUP TFI, COUP transcription factor 1, COUP transcription factor I, EAR 3, EAR3, ERBAL3, NR2F1, NR2F2, SVP44, TCFCOUP1, TFCOUP1, V erbA related protein 3
Gene Symbols NR2F1
Host Species Rabbit
Immunogen NR2F1 Fusion Protein Ag0118
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13865, 7025
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.