Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRF2/NFE2L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 50553534 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-553-534 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-553-534 Supplier Proteintech Group Inc Supplier No. 163961AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nrf2 (nuclear factor erythroid 2-related factor 2) is a transcription activator which plays a key role in protecting the cell from oxidative stress and inflammation. It is primarily localized in the cytoplasm under normal conditions, but under conditions of oxidative stress, Nrf2 migrates to the nucleus to upregulate expression of genes with ARE (antioxidant response elements) in the promoter region. The upregulated genes include those involved in cellular protection, drug transport, detoxification, and anti-oxidant defense. The protein is ubiquitously expressed in various tissues, with the highest levels found in kidney, muscle, lung, heart, liver, and brain.

Specifications

Antigen NRF2/NFE2L2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NFE2L2
Gene Accession No. O54968, Q16236, Q60795
Gene Alias HEBP1, NF E2 related factor 2, NFE2 related factor 2, NFE2L2, NRF2
Gene Symbols Nfe2l2
Host Species Rabbit
Immunogen NRF2, NFE2L2 Fusion Protein Ag9489
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18024, 4780, 83619
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.