Learn More
Invitrogen™ Nrf2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA138312
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is placenta or lung tissue.
Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.
Specifications
Nrf2 | |
Polyclonal | |
Unconjugated | |
NFE2L2 | |
AI194320; E4TF1-60; E4TF1A; HEBP1; Nfe2l2; NF-E2-related factor 2; NFE2-related factor 2; NF-E2-related factor-2; NFT2; NRF2; NRF2A; nuclear factor (erythroid-derived 2)-like 2; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2; Unknown (protein for IMAGE:7945830) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4780 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.21 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
Q16236 | |
NFE2L2 | |
Synthetic peptide corresponding to C-terminus of human Nrf2 protein. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.