Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nrf2 Recombinant Rabbit Monoclonal Antibody (HL1021)
GREENER_CHOICE

Catalog No. PIMA542371 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542371 Supplier Invitrogen™ Supplier No. MA542371
  • $617.50
    $504.65 / Each of 1

    Save $112.85

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nrf2
Applications Immunohistochemistry (Paraffin), Western Blot, Proximity Ligation Assay, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1021
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NFE2L2
Gene Accession No. O54968, Q16236, Q60795
Gene Alias AI194320; E4TF1-60; E4TF1A; HEBP1; Nfe2l2; NF-E2-related factor 2; NFE2-related factor 2; NF-E2-related factor-2; NFT2; NRF2; NRF2A; nuclear factor (erythroid-derived 2)-like 2; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2; Unknown (protein for IMAGE:7945830)
Gene Symbols NFE2L2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NRF2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18024, 4780, 83619
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.