Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRG1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2C6D12, Proteintech
SDP

Catalog No. 501736915 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6915 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6915 Supplier Proteintech Group Inc Supplier No. 664921IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The neuregulins are a family of ErbB/HER ligands encoded by four genes. Neuregulin-1 gene, NRG1, encodes numerous splice variants with differing transcription initiation sites. Neuregulin-1 includes a range of isoforms with varying glycosylation, regulation of expression and function. Neuregulin-1 splice variants each bear an EGF-like domain, though otherwise have unique domain structures, differing functions and discrete tissue distribution. Six types of Neuregulin-1 isoform groups have been defined based on their structural features. Three types are most often described, type I (ARIA, NDF or HRG), type II (GGF) and type III (SMDF). Neuregulin-1 has been linked to schizophrenia and has diverse neural functions. Neuregulin-1 affects cell migration, the differentiation of neural crest and Schwann cells, and acts to upregulate the expression of acetylcholine receptors at muscle fibers during the formation of neuromuscular junctions.

Specifications

Antigen NRG1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2C6D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NRG1
Gene Accession No. P43322, Q02297
Gene Alias ARIA, GGF, GGF2, Glial growth factor, Heregulin, HGL, HRG, HRG1, HRGA, NDF, Neu differentiation factor, neuregulin 1, NRG1, Pro NRG1, SMDF
Gene Symbols NRG1
Host Species Mouse
Immunogen NRG1 Fusion Protein Ag0803
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102164569, 112400, 211323, 3084
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.