Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599277 Supplier Invitrogen™ Supplier No. PA599277
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NRIP2.

NRIP2 down-regulates transcriptional activation by nuclear receptors such as NR1F2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRIP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NRIP2
Gene Accession No. Q9BQI9, Q9JHR9
Gene Alias AW491344; DKFZP761G1913; fj77f06; neuronal interacting factor 1; neuronal-interacting factor X 1; Nix1; Nrip2; nuclear receptor interacting protein 2; nuclear receptor interacting protein 2-like; nuclear receptor-interacting protein 2; si:dkey-245f7.1; similar to nuclear receptor interacting protein 2; wu:fj77f06
Gene Symbols NRIP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NRIP2(Accession Q9BQI9), corresponding to amino acid residues S34-R84.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60345, 83714
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.