Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRIP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599670
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599670 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599670 Supplier Invitrogen™ Supplier No. PA599670
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NRIP3.

NRIP3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRIP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Nrip3
Gene Accession No. Q9JJR9, Q9NQ35
Gene Alias A330103B05Rik; C11orf14; D7H11orf14; ICRFP703B1614Q5.2; ICRFP703N2430Q5.2; NRIP3; nuclear receptor interacting protein 3; nuclear receptor-interacting protein 3; NY-SAR-105; Sarcoma antigen NY-SAR-105
Gene Symbols Nrip3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NRIP3(Accession Q9NQ35), corresponding to amino acid residues L196-A241.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361625, 56675, 78593
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.