Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRXN1 Recombinant Rabbit Monoclonal Antibody (22H29L23)

Catalog No. 703650
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-365-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-365-0 Supplier Invitrogen™ Supplier No. 703650
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Cat, Sheep. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Neuronal cell surface protein that may be involved in cell recognition and cell adhesion by forming intracellular junctions through binding to neuroligins. May play a role in formation or maintenance of synaptic junctions. May mediate intracellular signaling. May play a role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRXN1
Applications Immunocytochemistry, Immunofluorescence
Classification Recombinant Monoclonal
Clone 22H29L23
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Nrxn1
Gene Accession No. Q9ULB1
Gene Alias 1700062G21Rik; 9330127H16Rik; A230068P09Rik; alpha-latrotoxin receptor (calcium-dependent); DKFZp313P2036; FLJ35941; Hs.22998; Kiaa0578; mKIAA0578; neurexin 1; Neurexin 1alpha; neurexin I; neurexin I-alpha; Neurexin I-beta; neurexin-1; Neurexin-1-alpha; neurexin-1-beta; non-processed neurexin I-alpha; Nrxn1; Nrxn1b; PTHSL2; SCZD17
Gene Symbols Nrxn1
Host Species Rabbit
Immunogen Peptides corresponding to human NRXN1 [aa296-aa313, aa394-aa406].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.