Learn More
Cygnus Technologies NS/0 HCP ELISA Kit, Successfully Qualified for a Number of Different Therapeutic Human Monoclonal Antibodies
Supplier:  Cygnus Technologies F220
Encompass
Hybridization of antibody producing cells with NS/0 cells is one of the most commonly employed systems for the production of humanized and mouse monoclonal antibodies. This kit has been successfully qualified for a number of different therapeutic human monoclonal antibodies. Our assay validation has
Catalog No. NC0199195
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.