Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NSC 23766

Catalog No. AAJ64979MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64979MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64979MA Supplier Thermo Scientific™ Supplier No. J64979MA

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

A cell-permeable pyrimidine compound that specifically and reversibly inhibits Rac1 GDP/GTP exchange activity.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable pyrimidine compound that specifically and reversibly inhibits Rac1 GDP/GTP exchange activity
CAS 733767-34-5
Molecular Formula C24H35N7·3HCl
Formula Weight 530.96g/mol
Synonym Rac1 Inhibitor
Sensitivity Light Sensitive and Hygroscopic
Product Type Signal Transduction Reagent
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.