Learn More
Abcam NSC 3852, Histone deacetylase inhibitor, 50MG
Supplier:  Abcam AB14588450MG
Encompass_Preferred
MW 174.16 g/mol, Purity >95%. Histone deacetylase (HDAC) inhibitor. Causes cell differentiation and antiproliferative activity in MCF-7 human breast cancer cells in vitro and displays antitumor activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5039
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.