Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSC 697923, Tocris Bioscience™

Catalog No. 485810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-581-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-581-0 Supplier R&D Systems Supplier No. 4858/10
This item is not returnable. View return policy

Selective UBE2N inhibitor

Selective inhibitor of the E2 ubiquitin (Ub) conjugating enzyme UBE2N (Ubc13). Inhibits UBE2N-Ub conjugation and activity of the UBE2N-Ueva1A complex. Has no effect on UbcH5c activity. Inhibits NF-κB activation in diffuse large B-cell lymphoma (DLBCL) cells. Inhibits cell growth and induces apoptosis in DLBCL and neuroblastoma (NB) cell lines. Supresses NB cell xenograft tumor growth in vivo.

Chemical Identifiers

CAS 343351-67-7
Molecular Formula C11H9NO5S
Molecular Weight (g/mol) 267.255
InChI Key GAUHIPWCDXOLCZ-UHFFFAOYSA-N
Synonym 2-4-methylphenyl sulfonyl-5-nitrofuran, 2-nitro-5-tosylfuran, 2-nitro-5-tosyl-furan, ncimech_000297, 2-nitro-5-p-toluenesulfonylfuran, 2-nitro-5-p-tolylsulfonyl furan, 2-nitro-5-toluene-4-sulfonyl-furan, 2-4-methylphenyl sulfonyl-5-nitro-furan, furan, 2-4-methylphenyl sulfonyl-5-nitro
PubChem CID 394449
IUPAC Name 2-(4-methylphenyl)sulfonyl-5-nitrofuran
SMILES CC1=CC=C(C=C1)S(=O)(=O)C2=CC=C(O2)[N+](=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 267.26
Percent Purity >98%
Chemical Name or Material NSC 697923
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.