Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSPL2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987892
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7892 Supplier Bioss Supplier No. BS7644R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.

Specifications

Antigen NSPL2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RTN3
Gene Accession No. O95197
Gene Alias ASY interacting protein; ASYIP; HAP; Homolog of ASY protein; Neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; NSPL2; NSPLII; Nsp-like II; NSP-like protein 2; NSP-like protein II; reticulon 3; Reticulon-3; Rtn3; RTN3-A1
Gene Symbols RTN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NSPL2/RTN3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10313
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.