Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSUN5P1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569876 Supplier Invitrogen™ Supplier No. PA569876
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 91%; Horse: 100%; Human: 100%; Mouse: 91%; Pig: 91%; Rat: 91%.

This locus represents a transcribed pseudogene of a nearby locus on chromosome 7, which encodes a putative methyltransferase. There is also a third closely related pseudogene locus in this region. Alternative splicing results in multiple transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSUN5P1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NSUN5P1
Gene Accession No. Q3KNT7
Gene Alias NOP2/Sun RNA methyltransferase family member 5 pseudogene 1; NSUN5B; NSUN5P1; WBSCR20B
Gene Symbols NSUN5P1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human NSUN5P1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155400
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.