Learn More
Description
Specifications
Specifications
| Calibration Range | 125 to 2000pg/mL |
| Conjugate | HRP |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Includes | One 96-well plate pre-coated with anti-NT-proBNP antibody (96 (8 x 12) wells), Standard: (2250pg/mL): 0.5mL, Wash buffer (30x): Dilution: 1:30 (20mL), Standard diluent buffer (1.5mL), Sample diluent buffer (6mL), HRP conjugated anti-NT-proBNP antibody (RTU): 6mL, Stop solution (6mL), TMB chromogenic reagent A (6mL), TMB chromogenic reagent B (6mL), Plate sealer (2 slices), One hermetic bag |
| Regulatory Status | For research use only (RUO) |
| Sample Type | CCS, Plasma, Serum, Tissue, Urine |
| Sample Volume | 50 μL |
| Content And Storage | 4°C. Avoid prolonged exposure to light |
| For Use With (Application) | Sandwich enzyme immunoassay for the quantitative measurement of human N-terminal prohormone of brain natriuretic peptide (NT-proBNP) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.