Learn More
Advanced ImmunoChemical, Inc. NT-PROBNP, HUMAN RECOMBINANT
Supplier:  Advanced ImmunoChemical, Inc. 8NTPBRH0.1 MG
Encompass_Preferred
ANTIGENS/Source:Recombinant E.coli.Applications:Calibrator or standard in immunoassays, immunogen for antibody production.Analysis:Purity > 95 % (Tricine-SDS-PAGE).Concentration is determined spectrophotometrically.Immunoreactivity is confirmed by reaction with monoclonal antibodies specific to different fragments of NT-proBNP molecule.Predicted molecular mass 8589 Da confirmed by MALDI-MS.Recombinant human NT-proBNP contains additional methionine residue at the N-terminus (in comparison with native NT-proBNP).Purification:Immunoaffinity purification followed by ion-exchange chromatography.Presentation:10 mM potassium phosphate, pH 7.4, 150 mM NaClStorage Conditions:-70 °C (between -65 and -80 °C allowed)
Catalog No. NC3566233
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.