Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5DC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570919 Supplier Invitrogen™ Supplier No. PA570919
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 86%; Zebrafish: 77%.

NT5DC3 gene ontology annotations related to this gene include 5'-nucleotidase activity; metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5DC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NT5DC3
Gene Accession No. Q86UY8
Gene Alias 5'-nucleotidase domain containing 3; 5'-nucleotidase domain-containing protein 3; AU040402; AW540062; C630002B14Rik; GNN; GRP94-neighboring nucleotidase; NT5DC3; TU12B1TY; TU12B1-TY; TU12B1-TY protein homolog
Gene Symbols NT5DC3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human NT5DC3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51559
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.