Learn More
Cytiva NTA REAGENT KIT
Supplier:  Cytiva 28995043
Encompass
Reagents for Sensor Chip NTA, which is used to capture histidine-tagged molecules in biomolecular interaction analysis.
Catalog No. 50-105-5520