Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01676757 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-757 Supplier Invitrogen™ Supplier No. PA119441
  • $510.00
    $425.65 / Each of 1

    Save $84.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody recognizes N-terminal cytoplasmic domain of mouse Non-T cell Activation Linker (NTAL), also known as LAB (linker of activated B cells), a 25 kDa transmembrane adaptor protein present in membrane microdomains (rafts) of B cells, NK cells and myeloid cells. Western Blot: non-reducing conditions.

NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTAL
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene LAT2
Gene Accession No. Q9JHL0
Gene Alias AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5
Gene Symbols LAT2
Host Species Rabbit
Immunogen Recombinant N-terminal cytoplasmic domain (aa 124-203) of mouse NTAL.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56743
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.