Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTCP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580001 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580001 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580001 Supplier Invitrogen™ Supplier No. PA580001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, mouse liver tissue. IHC: mouse liver tissue, rat liver tissue IHC-F: mouse liver tissue. Flow: RAW264.7 cell.

Sodium/bile acid cotransporters are integral membrane glycoproteins that participate in the enterohepatic circulation of bile acids. Two homologous transporters are involved in the reabsorption of bile acids, one absorbing from the intestinal lumen, the bile duct, and the kidney with an apical localization (SLC10A2), and the other being found in the basolateral membranes of hepatocytes (SLC10A1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTCP
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene SLC10A1
Gene Accession No. O08705, P26435
Gene Alias bile acid cotransporting polypeptide; cell growth-inhibiting gene 29 protein; GIG29; growth-inhibiting protein 29; hepatic sodium-dependent bile acid transporter; LOW QUALITY PROTEIN: sodium/bile acid cotransporter; MGC128766 protein; Na(+)/bile acid cotransporter; Na(+)/taurocholate transport protein; Na/taurocholate cotransporting polypeptide; NA-dependent cholate transporting protein; Ntcp; Ntcp1; SBACT; Slc10a1; sodium bile acid cotransporting polypeptide; sodium/bile acid cotransporter; sodium/tauro; sodium/taurocholate cotransporter; sodium/taurocholate cotransporting polypeptide; sodium-dependent bile acid cotransporter; sodium-dependent taurocholate cotransporting polypeptide; sodium-taurocholate cotransporting polypeptide; solute carrier family 10 (sodium/bile acid cotransporter family), member 1; solute carrier family 10 (sodium/bile acid cotransporter), member 1; solute carrier family 10 member 1; solute carrier family 10, member 1
Gene Symbols SLC10A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of mouse SLC10A1 (296-336aa EGLLFIIIFRCYLKIKPQKDQTKITYKAAATEDATPAALEK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20493, 24777
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.