Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101720 Supplier Invitrogen™ Supplier No. PA5101720
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PNPLA6.

This gene encodes a phospholipase that deacetylates intracellular phosphatidylcholine to produce glycerophosphocholine. It is thought to function in neurite outgrowth and process elongation during neuronal differentiation. The protein is anchored to the cytoplasmic face of the endoplasmic reticulum in both neurons and non-neuronal cells. Mutations in this gene result in autosomal recessive spastic paraplegia, and the protein is the target for neurodegeneration induced by organophosphorus compounds and chemical warfare agents. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTE
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene pnpla6
Gene Accession No. Q3TRM4, Q8IY17
Gene Alias 5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI661849; BNHS; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; iPLA2delta; LNMS; MSws; Neuropathy target esterase; NT; NT5; NT5E; NTE; NTEMND; OMCS; patatin like phospholipase domain containing 6; patatin-like phospholipase domain containing 6; patatin-like phospholipase domain-containing protein 6; PNPLA6; Purine 5-Prime-Nucleotidase; RGD1564611; SPG39; Swiss cheese; Swiss-cheese; sws; wu:fy20f06
Gene Symbols pnpla6
Host Species Rabbit
Immunogen A synthesized peptide derived from human PNPLA6(Accession Q8IY17), corresponding to amino acid residues L326-R376.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10908, 50767
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.