Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670635
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-635 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-635 Supplier Invitrogen™ Supplier No. PA118383
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute in 500 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTF3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ntf3
Gene Accession No. P18280, P20783, P25433
Gene Alias AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3
Gene Symbols Ntf3
Host Species Sheep
Immunogen Synthetic peptide corresponding to residues Y(139) A E H K S H R G E Y(149) of human, mouse and rat NT3.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 428099, 4908, 81737
Target Species Human, Rat, Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.