Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674406 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-406 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-406 Supplier Invitrogen™ Supplier No. PA19523
  • $366.00
    $303.65 / Each of 1

    Save $62.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chicken Polyclonal Antibody

PA1-9523 detects NT4/5 in human and mouse samples. PA1-9523 has been successfully used in ELISA procedures. PA1-9523 immunogen is a synthetic peptide based on the human NT4/5 protein.

This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTF4
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NTF4
Gene Accession No. P34130, Q80VU4
Gene Alias 2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5
Gene Symbols NTF4, Ntf5
Host Species Chicken
Immunogen Synthetic peptide mixture based on the human NT4/5 protein.
Purification Method IgY fraction
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4909, 78405
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.