Learn More
AdipoGen Nitrilotri(methylphosphonic acid) (10 g)
SDP
Supplier:  AdipoGen CDXN0256G010
Encompass_Preferred
Chemical. CAS: 6419-19-8. Formula: C3H12NO9P3. MW: 299.05. Synthetic. Building block and powerful complexing agent. Nitrilotri(methylphosphonic acid) is a common chelating agent used in synthetic chemistry. It is used for the preparation of hexagonal porous three-dimensional structures encapsulating a template, layered structures with intercalated templates or linear polymers, for the synthesis of metal-organic frameworks in combination with uranyl nitrate or the preparation of anticorrosive protective coatings. ATMP can also be employed as an anti-fouling agent (scale inhibitor). It can prevent scale formation in water systems. It is used as detergents and cleaning agent, water treatment and antiscale agent.
Catalog No. 50-205-4560
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.