Learn More
Medchemexpress LLC 2-({[1-(4-fluorophenyl)-5-(2-methoxyphenyl)-1h-pyrazol-3-yl]carbonyl}amino)tricyclo[3.3.1.13,7]decane-2-... | 1811503-67-9 | 95.6% | 489.54 g/mol | C28H28FN3O4 | 50MG
Supplier: Medchemexpress LLC HY12483150MG
NTRC-844 is a small-molecule research compound that acts as a selective antagonist of neurotensin receptor type 2 (NTS2). It has an EC50 of 238 nM and is used in preclinical analgesia and pharmacology studies; analytical characterization and safety documentation are available.