Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675194
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-194 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-194 Supplier Invitrogen™ Supplier No. PA141575
  • $603.00
    $477.20 / Each of 1

    Save $125.80

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: brain lysate, human brain protein. Predicted Reactivity: Rat, Dog, Bovine and Mouse.

The protein encoded by this gene belongs to the G protein-coupled receptor family that activate a phosphatidylinositol-calcium second messenger system. Binding and pharmacological studies demonstrate that this receptor binds neurotensin as well as several other ligands already described for neurotensin NT1 receptor. However, unlike NT1 receptor, this gene recognizes, with high affinity, levocabastine, a histamine H1 receptor antagonist previously shown to compete with neurotensin for low-affinity binding sites in brain. These activities suggest that this receptor may be of physiological importance and that a natural agonist for the receptor may exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTSR2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene NTSR2
Gene Accession No. O95665, P70310
Gene Alias High-affinity levocabastine-sensitive neurotensin receptor; levocabastine-sensitive neurotensin receptor; low affinity neurotensin receptor; low-affinity levocabastine-sensitive neurotensin receptor; neurotensin receptor 2; neurotensin receptor type 2; neurotensin receptor, type 2; NT2 receptor; NT2R; NTR2; NT-R-2; NTR2 receptor; NTRL; Ntsr2
Gene Symbols NTSR2
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human Neurotensin receptor 2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18217, 23620
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.