Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NucA nuclease Antibody (1028905) [HRP], R&D Systems™
SDP

Catalog No. FAB100633H
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB100633H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB100633H Supplier R&D Systems Supplier No. FAB100633H

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

NucA nuclease Monoclonal specifically detects NucA nuclease in Multi-species samples. It is validated for Western Blot.

Specifications

Antigen NucA nuclease
Applications Western Blot
Classification Monoclonal
Clone 1028905
Conjugate HRP
Formulation PBS
Gene Alias DcD
Host Species Mouse
Immunogen E. coli -derived s. marcescens NucA nuclease, Met1-Asn266, Accession # P13717
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233874471
Target Species Multi-species
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.