Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nuclear Antigen Monoclonal Antibody (235-1)
GREENER_CHOICE

Catalog No. PIMA533098 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533098 Supplier Invitrogen™ Supplier No. MA533098
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells, including neurons and embryonic stem cells, with no reactivity to rat or mouse cells. This clone stains nuclei of all human cell types giving a diffuse nuclear staining pattern; chromosomes are negatively stained in metaphase cells. With immunoprecipitation, the antibody reacts with a ~80 kDa and ~70 kDa band. The antibody works on acetone-fixed and paraformaldehyde-fixed cells. It is suggested to permeabilize the cells with 0.1-0.4% Triton X-100. The antibody also works for immunohistochemistry on paraformaldehyde fixed cryostat sections.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Antigen
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 235-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Tissue culture supernatant with no preservative
Gene Alias Histone H1; HNA; HuNu; Nuclei; Nucleus
Host Species Mouse
Immunogen Human cell homogenate.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.